M35355
Glutaric acid monomethyl ester chloride
98%
Synonym(s):
Methyl 4-(chloroformyl)butyrate, Methyl 5-chloro-5-oxovalerate, Methyl glutaryl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ClCO(CH2)3COOCH3
CAS Number:
Molecular Weight:
164.59
Beilstein:
1762383
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.445 (lit.)
bp
110 °C/17 mmHg (lit.)
density
1.191 g/mL at 25 °C (lit.)
SMILES string
COC(=O)CCCC(Cl)=O
InChI
1S/C6H9ClO3/c1-10-6(9)4-2-3-5(7)8/h2-4H2,1H3
InChI key
JCAZSWWHFJVFPP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
179.6 °F - closed cup
Flash Point(C)
82 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service