Skip to Content
Merck
All Photos(1)

Key Documents

CLS31666

Pyrex® cloning cylinder

O.D. × H 6 mm × 8 mm

Synonym(s):

lab equipment accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106618
NACRES:
NB.43

material

borosilicate glass
cylindrical

sterility

non-sterile

packaging

pack of 125

manufacturer/tradename

Corning 3166-6

O.D. × H

6 mm × 8 mm

Looking for similar products? Visit Product Comparison Guide

General description

Clone a single cell or group of cells by surrounding them with this Pyrex glass open ended cloning cylinder. Dip the end of the cylinder into sterile silicone grease before pressing to the bottom of a culture dish to create an isolated well. Cylinders are autoclavable or can be dry heat sterilized.

Features and Benefits

  • Pyrex® cloning cylinder is made of Type I Class A - Low-Expansion Borosilicate Glass.
  • Compatible with heating using a hot plate, flame, oven, microwave or heating mantles.
  • Product can be cooled to -230°C.
  • Product Dimensions - 6 mm OD x 8 mm High.
  • DO NOT plunge product directly into a liquid coolant.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service