BAH367140000
SP Bel-Art® Quaver Vibrating Spatula
stainless steel blade
Synonym(s):
spoon
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
Recommended Products
material
stainless steel blade
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H36714-0000
Related Categories
General description
Make accurate additions of precious materials as needed. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving you control over transfer from spatula to weighing dish.
Features and Benefits
- Available with a plain stainless steel blade or with a brown FEP coating
- Coating complies with FDA regulations governing direct food contact applications; reduces transfer losses
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP