BAF430000001
Shelf set for Scienceware® Grande desiccator
acrylic, perforated, pack of 2 ea
Synonym(s):
Scienceware, dry-box desiccator cabinet, low humidity
All Photos(1)
About This Item
Recommended Products
feature
acrylic, perforated
packaging
pack of 2 ea
manufacturer/tradename
Bel-Art F43000-0001
General description
Grande Desiccator can hold up to seven shelves spaced 2 inches apart, pack of 2 (two are included with desiccator).
The desiccant used in the cartridge is: Z163414
- Pull-out, perforated acrylic shelves allow maximum air circulation and have built-in stops to prevent accidental tipping; maximum weight per shelf: 10 lbs (4.5 kg)
- Shelves are removable to allow use as a single, spacious chamber for specific storage needs
- Dimensions: 50.8 W x 53 D x 6.7 cm H (20 x 20 7/8 x 2 5/8 in.
The desiccant used in the cartridge is: Z163414
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)