Skip to Content
Merck
All Photos(1)

Key Documents

922757

Sigma-Aldrich

(4-Chlorophenyl)(5,5-dimethyl-2-oxido-1,3,2-dioxaphosphinan-2-yl)methanone

≥95%

Synonym(s):

p-chlorobenzoylphosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14ClO4P
CAS Number:
Molecular Weight:
288.66
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

105-107 °C

functional group

chloro
phosphonate

storage temp.

−20°C

Application

This acylphosphonate can be used as a reagent in the cobalt catalyzed radical hydroacylation of alkenes to ketones. This reagent can be used under mild conditions, and is tolerant of various functional groups, making it a versatile reagent for the installation of ketones into a variety of compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service