All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C7H7BN4O2
CAS Number:
Molecular Weight:
189.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95%
form
solid
mp
148-152 °C
storage temp.
2-8°C
SMILES string
OB(O)c1ccccc1-c2nnn[nH]2
InChI
1S/C7H7BN4O2/c13-8(14)6-4-2-1-3-5(6)7-9-11-12-10-7/h1-4,13-14H,(H,9,10,11,12)
InChI key
GVRXWYFECKHTSJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2-(Tetrazol-5-yl)phenylboronic acid can be used as a reactant to prepare:
- Biaryltetrazole derivatives via Suzuki-Miyaura cross-coupling with aryl and heteroaryl halides via formation of C-C bond.
- 2-Aminobenzoxazole derivatives containing biphenyl tetrazole as potent human chemR23 inhibitors.
- o-Phenylenediamine possessing tetrazolyl derivatives as potent inhibitors of indoleamine 2,3-dioxygenase.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The discovery and optimization of a series of 2-aminobenzoxazole derivatives as ChemR23 inhibitors
Imaizumi T, et al.
Bioorganic & Medicinal Chemistry, 27(21), 115091-115091 (2019)
Development of a series of novel o-phenylenediamine-based indoleamine 2, 3-dioxygenase 1 (IDO1) inhibitors
Williams DK, et al.
Bioorganic & Medicinal Chemistry Letters, 28(4), 732-736 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service