Skip to Content
Merck
All Photos(1)

Key Documents

684686

Sigma-Aldrich

2,2-Difluoropropionic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4F2O2
CAS Number:
Molecular Weight:
110.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

powder

bp

25-30 °C/5 mmHg

mp

38-45 °C

functional group

carboxylic acid
fluoro

SMILES string

CC(F)(F)C(O)=O

InChI

1S/C3H4F2O2/c1-3(4,5)2(6)7/h1H3,(H,6,7)

InChI key

PMWGIVRHUIAIII-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.1 °F - closed cup

Flash Point(C)

65.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Morales-Cerrada et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(1), 296-308 (2018-09-20)
Thermal decarbonylation of the acyl compounds [Mn(CO)5 (CORF )] (RF =CF3 , CHF2 , CH2 CF3 , CF2 CH3 ) yielded the corresponding alkyl derivatives [Mn(CO)5 (RF )], some of which have not been previously reported. The compounds were fully

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service