Skip to Content
Merck
All Photos(1)

Documents

672343

Sigma-Aldrich

2-(Di-tert-butylphosphino)-1-phenylindole

95%

Synonym(s):

N-Phenyl-2-(di-tert-butylphosphino)indole, N-Phenylindol-2-yl-di-tert-butylphosphine, cataCXium® PIntB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28NP
CAS Number:
Molecular Weight:
337.44
Beilstein:
9924511
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reaction type: Asymmetric synthesis
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

functional group

phosphine

SMILES string

CC(C)(C)P(c1cc2ccccc2n1-c3ccccc3)C(C)(C)C

InChI

1S/C22H28NP/c1-21(2,3)24(22(4,5)6)20-16-17-12-10-11-15-19(17)23(20)18-13-8-7-9-14-18/h7-16H,1-6H3

InChI key

HDZRDZCQFYUOHE-UHFFFAOYSA-N

General description

sold in collaboration with Solvias AG

Legal Information

WO 2004/101581
cataCXium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service