Skip to Content
Merck
All Photos(1)

Documents

60420

Sigma-Aldrich

Potassium oleate

technical, ≥87% (fatty acid)

Synonym(s):

Oleic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOK
CAS Number:
Molecular Weight:
320.55
Beilstein:
4167152
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥87% (fatty acid)

impurities

≤0.3% free KOH
≤1.5% humidity

SMILES string

[K+].[H]\C(CCCCCCCC)=C(/[H])CCCCCCCC([O-])=O

InChI

1S/C18H34O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h9-10H,2-8,11-17H2,1H3,(H,19,20);/q;+1/p-1/b10-9-;

InChI key

MLICVSDCCDDWMD-KVVVOXFISA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium oleate can be used to synthesize uncoagulative oleic acid magnetic ultrafine particles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Preparation of ultrafine particle multilayers using the Langmuir?Blodgett technique"
<BIG>Nakaya T, et al.</BIG>
Journal of Materials Chemistry, 6(05), 691-697 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service