Skip to Content
Merck
All Photos(1)

Key Documents

237655

Sigma-Aldrich

(−)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane

98%

Synonym(s):

(−)-1,4-Bis(diphenylphosphino)-1,4-dideoxy-2,3-O-isopropylidene-L-threitol, (4R,5R)-4,5-Bis(diphenylphosphino-methyl)-2,2-dimethyl-1,3-dioxolane, (4R-trans)-[(2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene)]bis(diphenylphosphine), (R,R)-DIOP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H32O2P2
CAS Number:
Molecular Weight:
498.53
Beilstein:
1441045
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

optical activity

[α]19/D −26°, c = 2.3 in chloroform

mp

88-90 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)O[C@@H](CP(c2ccccc2)c3ccccc3)[C@H](CP(c4ccccc4)c5ccccc5)O1

InChI

1S/C31H32O2P2/c1-31(2)32-29(23-34(25-15-7-3-8-16-25)26-17-9-4-10-18-26)30(33-31)24-35(27-19-11-5-12-20-27)28-21-13-6-14-22-28/h3-22,29-30H,23-24H2,1-2H3/t29-,30-/m0/s1

InChI key

VCHDBLPQYJAQSQ-KYJUHHDHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane is a C2 chelating diphosphine ligand for transition metal complexes during asymmetric catalysis.

Application

For the in situ preparation of chiral hydrogenation catalysts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(2, 3-O-Isopropylidene)-2, 3-dihydroxy-1, 4-bis (diphenylphosphino) butane
Kagan H and Anderson MB
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service