Skip to Content
Merck
All Photos(1)

Key Documents

222259

Sigma-Aldrich

2-Methyl-5-phenylbenzoxazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11NO
CAS Number:
Molecular Weight:
209.24
Beilstein:
159076
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

60-64 °C (lit.)

functional group

phenyl

SMILES string

Cc1nc2cc(ccc2o1)-c3ccccc3

InChI

1S/C14H11NO/c1-10-15-13-9-12(7-8-14(13)16-10)11-5-3-2-4-6-11/h2-9H,1H3

InChI key

CHZDPBVCBBQQRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-5-phenylbenzoxazole was used in the preparation of 2-methyl-3-ethyl-5-phenyl-1H-benzoxazolium iodide (QM1).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mobasher Ahmad
Scientia pharmaceutica, 79(4), 921-935 (2011-12-07)
The relative lack of clinical success with conventional anticancer agents may be due in part to the traditional concept of cancer being a biological state rather than a dynamic process. Redefining cancer as a dynamic disease commencing with carcinogenesis introduces
Peng ZH, et al.
Journal of Materials Chemistry, 6(8), 1325-1333 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service