ME14-1KT
Fatty Acid Methyl Esters, Unsaturated Kit
(individually packaged in quantities indicated), analytical standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
85151701
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
CofA
current certificate can be downloaded
packaging
pkg of 1 kit
concentration
(individually packaged in quantities indicated)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
food and beverages
format
neat
functional group
carboxylic acid
ester
shipped in
dry ice
storage temp.
−20°C
Related Categories
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Analyte
Description
cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester 100 mg
Methyl arachidonate 100 mg
Methyl cis-13-docosenoate 100 mg
Methyl cis-11-eicosenoate 100 mg
Methyl elaidate 100 mg
Methyl linoleate 100 mg
Methyl linolelaidate 100 mg
Methyl linolenate 100 mg
Methyl myristoleate 100 mg
Methyl oleate 100 mg
Methyl palmitoleate 100 mg
Methyl petroselinate 100 mg
Methyl cis-15-tetracosenoate 100 mg
See All (13)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Elena Cartea et al.
Foods (Basel, Switzerland), 8(8) (2019-07-31)
The seed oil content and the fatty acid composition of a germplasm collection of Brassica napus and Brassica rapa currently grown in Galicia (northwestern Spain) were evaluated in order to identify potentially interesting genotypes and to assess their suitability as
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service