Skip to Content
Merck
All Photos(1)

Key Documents

82037-U

Hamilton® Hydrogen Form (12 μm) HPLC Columns

L × I.D. 2 mm × 4.6 mm, stainless steel hardware, HPLC Guard Cartridge Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Hamilton Hydrogen Form HPLC Guard Cartridge, Kit, 12 μm particle size, L × I.D. 2 mm × 4.6 mm, stainless steel hardware

material

stainless steel hardware

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

2 mm × 4.6 mm

matrix

Hydrogen Form
polymeric

particle size

12 μm

operating pH

1-14

separation technique

ion exclusion

Looking for similar products? Visit Product Comparison Guide

General description

Kit includes 1 holder and 2 cartridges

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service