Skip to Content
Merck
All Photos(1)

Key Documents

814477

TOYOPEARL® Phenyl-650S Bulk Media

phase phenyl, bottle of 100 mL, 35 μm particle size

Synonym(s):

TOYOPEARL® Phenyl-650 Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
particle size:
35 μm
matrix active group:
phenyl phase
matrix:
polymer (35 μm)

description

Phenyl-650S

product line

TOYOPEARL®

form

slurry

packaging

bottle of 100 mL

parameter

≤50% organic solvent
3 bar max. pressure

technique(s)

LPLC: suitable

matrix

polymer (35 μm)

matrix active group

phenyl phase

particle size

35 μm

operating pH

1-13

separation technique

hydrophobic interaction (HIC)

General description

Toyopearl Phenyl-650 resin is the middle hydrophobicity of the five HIC ligands offered by Tosoh Bioscience. Primary applications are for the separation of proteins, their isoforms, and aggregate removal. It is typically used in intermediate purification and polishing steps. For high performance separations, the use of TSKgel Phenyl-5PW resin is suggested. Both products have the same selectivity.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service