Z719595
Sorenson™ low binding standard tips
maximum volume 200 μL, pkg of 96 tips CellRack, sterile
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
200 μL
sterility:
sterile
feature:
barrier: no
packaging:
case of 4800 ea (50 racks of 96 tips)
pkg of 96 tips CellRack
pack of 960 ea (10 racks of 96 tips)
pkg of 96 tips CellRack
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Sorenson 15280T
Recommended Products
material
colorless
natural tip
polypropylene
sterility
sterile
feature
barrier: no
packaging
case of 4800 ea (50 racks of 96 tips)
pkg of 96 tips CellRack
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sorenson 15280T
maximum volume
200 μL
volume
200 μL
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)