SYNB141000
Synthware™ Glass wide mouth media bottle
borosilicate glass 3.3, capacity 1000 mL, red polypropylene cap
Synonym(s):
Synthware B141000, glass graduated bottle with screw cap
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Recommended Products
material
borosilicate glass 3.3
clear bottle
red polypropylene cap
wide-mouth
sterility
non-sterile
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
Kemtech B140100
capacity
1000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
GL45 Wide Mouth Media Storage Bottle With Color Caps, 1000mL includes Accessories: GL45-C
Application
kemetch; synthware; media bottle; GL45
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service