Skip to Content
Merck
All Photos(1)

Key Documents

SRP3090

Sigma-Aldrich

IL-3 human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

HCGF, MCGF (Mast cell growth factor), Multi-CSF, P-cell stimulation factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.1 ng/mL ED50

mol wt

15.0 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL3(3562)

General description

IL3 (interleukin 3) is a haematopoietic growth factor, the gene for which is localized to human chromosome locus 5.[1]
Recombinant human IL-3 is a 15.0 kDa globular protein containing 133 amino acid residues.

Biochem/physiol Actions

IL3 (interleukin 3) promotes the survival, proliferation and differentiation of haematopoietic progenitor cells. It also functions as an activator of basophils thus, resulting in significant increase in antigen and IgE-mediated release of cytokines and other mediators. Certain variants in this gene confer protection against malaria attack, and thus, this gene might play a role in the pathophysiology of falciparum malaria.[1] SNP (single nucleotide polymorphism) rs40401 in this gene and smoking have a positive correlation with asthma in Japanese women.[2] IL3 is involved in brain development and has a direct influence on the proliferation and survival of neural progenitor cell population, thereby also controlling human brain volume.[3]

Sequence

APMTQTTSLK TSWVNCSNMI DEIITHLKQP PLPLLDFNNL NGEDQDILME NNLRRPNLEA FNRAVKSLQN ASAIESILKN LLPCLPLATA APTRHPIHIK DGDWNEFRRK LTFYLKTLEN AQAQQTTLSL AIF

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian G Meyer et al.
Human molecular genetics, 20(6), 1173-1181 (2011-01-13)
Using segregation analyses, control of malaria parasites has previously been linked to a major gene within the chromosomal region 5q31-33, but also to complex genetic factors in which effects are under substantial age-dependent influence. However, the responsible gene variants have
Y Miyake et al.
Scandinavian journal of immunology, 79(6), 410-414 (2014-04-02)
Three previous genetic association studies conducted in Korea and China investigated the relationship between IL3 single nucleotide polymorphism (SNP) rs40401 and asthma, but their results were inconsistent. We examined this relationship and the possibility of an interaction between IL3 SNP
Xiong-jian Luo et al.
PloS one, 7(11), e50375-e50375 (2012-12-12)
One of the most significant evolutionary changes underlying the highly developed cognitive abilities of humans is the greatly enlarged brain volume. In addition to being far greater than in most other species, the volume of the human brain exhibits extensive
J N Ihle
Chemical immunology, 51, 65-106 (1992-01-01)
Considerable information has accumulated over the past several years relating to the biological activities of IL-3 which supports that concept that IL-3 has a unique capacity to support the proliferation and differentiation of a wide spectrum of hematopoietic cells. The
S Yousefi et al.
Allergy, 70(9), 1184-1188 (2015-06-05)
Basophils are primarily associated with immunomodulatory functions in allergic diseases and parasitic infections. Recently, it has been demonstrated that both activated human and mouse basophils can form extracellular DNA traps (BETs) containing mitochondrial DNA and granule proteins. In this report

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service