Skip to Content
Merck
All Photos(1)

Key Documents

SML3488

Sigma-Aldrich

CuATSP

≥98% (HPLC)

Synonym(s):

(SP-4-2)-[[2,2′-(1,2-Dimethyl-1,2-ethanediylidene)bis[N-phenylhydrazinecarbothioamidato-κN2,κS]](2-)]copper, Cu ATSP, Cu-ATSP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18CuN6S2
CAS Number:
Molecular Weight:
446.05
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, Faint brown to dark red

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

CC1=[N]2[Cu]3(SC(NC4=CC=CC=C4)=N2)SC(NC5=CC=CC=C5)=N[N]3=C1C

Biochem/physiol Actions

CuATSM analog with >10-fold enhanced ferropotosis inhibitory potency.
CuATSP is a CuATSM analog with >10-fold enhanced ferropotosis inhibitory potency (CuATSP/CuATSM IC50 against 150 nM RSL3-induced ferroptosis = 8.5/160 nM using Pfa1 mouse embryonic fibroblasts,16/170 nM using HT-22 mouse hippocampal cells). CuATSP and CuATSM exert their radical-trapping antioxidant (RTA) and (phospho)lipid peroxidation inhibitory potency via an initial reversible addition of a peroxyl radical to the bis(thiosemicarbazone) ligand, followed by a subsequent H-atom transfer (HAT) that drives the reaction forward.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radical-Trapping Antioxidant Activity of Copper and Nickel Bis(Thiosemicarbazone) Complexes Underlies Their Potency as Inhibitors of Ferroptotic Cell Death.
Zilka O, Poon JF, Pratt DA
Journal of the American Chemical Society, 143, 19043-19057 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service