Skip to Content
Merck
All Photos(1)

Key Documents

SML1822

Sigma-Aldrich

Akebia saponin D

≥98% (HPLC)

Synonym(s):

3-O-α-L-arabinopyranosyl hederagenin-28-β-D-glucopyranoside-(1→6)-β-D-glucopyranoside, ASD, Asperosaponin VI, Leiyemudanoside A, Tauroside St-G0-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H76O18
CAS Number:
Molecular Weight:
929.10
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

(Dipsacus asper)

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +13 to +18°, c = 0.5 in methanol

storage condition

desiccated

color

white to beige

solubility

H2O: 3 mg/mL, clear (warmed)

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC1(C)CC[C@]2(C(O[C@@H]3O[C@H](CO[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)=O)CC[C@@]5(C)[C@]6(C)CCC7[C@](C)(CO)[C@@H](O[C@]8([H])OC[C@H](O)[C@H](O)[C@H]8O)CC[C@]7(C)[C@@]6([H])CC=C5[C@]2([H])C1

InChI

1S/C47H76O18/c1-42(2)13-15-47(41(59)65-40-37(58)34(55)32(53)26(63-40)20-61-38-36(57)33(54)31(52)25(18-48)62-38)16-14-45(5)22(23(47)17-42)7-8-28-43(3)11-10-29(64-39-35(56)30(51)24(50)19-60-39)44(4,21-49)27(43)9-12-46(28,45)6/h7,23-40,48-58H,8-21H2,1-6H3/t23-,24-,25+,26+,27+,28+,29-,30-,31+,32+,33-,34-,35+,36+,37+,38+,39-,40-,43-,44-,45+,46+,47-/m0/s1

InChI key

CCRXMHCQWYVXTE-HMRSNRLKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Akebia saponin D (ASD) is a bioactive triterpenoid saponin isolated from the rhizome of Dipsacus asper Wall that is used as an anti-osteoporosis drug.
Akebia saponin D (ASD) is a bioactive triterpenoid saponin isolated from the rhizome of Dipsacus asper Wall that is used as an anti-osteoporosis drug. Akebia saponin D exhibits therapeutic effects in number of disease models including cancer, Alzheimer′s disease, cardiovascular disease, and bone fractures. Akebia saponin D protects against nonalcoholic fatty liver disease (NAFLD) liver damage in mice model of NAFLD. ASD decreases hepatic steatosis and heptocyte apoptosis through autophagy modulation. Akebia saponin D prevents oleic acid induced lipid droplets accumulation and increases autophagic flux BRL cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Li Gong et al.
The Journal of pharmacology and experimental therapeutics, 359(3), 392-400 (2016-10-27)
Nonalcoholic fatty liver disease (NAFLD) is considered to be a hepatic manifestation of the metabolic syndrome, and the incidence of NAFLD is increasing rapidly. However, appropriate drugs for treatment of NAFLD are lacking. This study aimed to elucidate the protective
Xing Yu et al.
Behavioural brain research, 235(2), 200-209 (2012-09-12)
Neuroinflammatory responses caused by amyloid β(Aβ) play an important role in the pathogenesis of Alzheimer's disease (AD). Aβ is known to be directly responsible for the activation of glial cells and induction of apoptosis. Akebia Saponin D (ASD) is extracted
Li-li Gong et al.
Journal of pharmacological sciences, 126(3), 243-252 (2014-10-17)
Akebia saponin D (ASD) is a typical bioactive triterpenoid saponin obtained from the rhizome of Dipsacus asper Wall. Previous studies have found that ASD has a hepatoprotective effect in a mouse model. The purpose of this paper was to explore

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service