GE28-9355-98
Dextrin Sepharose™ High Performance
Cytiva 28-9355-98, pack of 100 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106500
NACRES:
NA.56
Recommended Products
ligand
dextrin
packaging
pack of 100 mL
manufacturer/tradename
Cytiva 28-9355-98
storage condition
(20% Ehtanol)
matrix
highly cross-linked 6% agarose
avg. part. size
34 μm
cleaning in place
2-13
working range
>7
capacity
~7 mg binding capacity (MBP- ΔSal/ml medium Approx. 16 mg MBP- ßGal/ml medium)
storage temp.
2-8°C
Related Categories
General description
Binding capacity is protein dependent. Dynamic binding capacity was tested with MBP-?Sal (MBP2*-paramyosin-?Sal, Mr approx. 70 000, multimer in solution), and MBP-ßGal (MBP2*-beta-galactosidase, Mr approx. 158 000, multimer in solution).
Dextrin Sepharose™ High Perfromance is an affinity media for purification of proteins tagged with Maltose Binding protein (MBP).
Application
Dextrin Sepharose™ High Performance and provide convenient affinity purification of recombinant Proteins tagged with maltose binding Protein (MBP). This robust and stable Sepharose™ High Performance based medium enables elution of target Protein in narrow peaks, which minimizes the need for further concentration steps.
Features and Benefits
- Dextrin Sepharose™ High Performance is a robust, high resolution medium based on the 34 μm Sepharose™ High Performance matrix
- The small, evenly sized beads ensure that MBP-tagged Proteins elute in narrow peaks, thus minimizing the need for further concentration steps
- Physiological conditions and mild elution preserve target Protein activity
- Dextrin Sepharose™ High Performance tolerates all commonly used aqueous buffers and is easily regenerated using 0.5 M NaOH allowing the same column to be used for repeated purifications
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Sepharose is a trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service