Skip to Content
Merck
All Photos(1)

Key Documents

EMU016691

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Fis1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGAGACTGTGGCCCAGTAGAGACCTTAGTGTGAGGCTTTCAGGGGCGGCGGCCATGGAGGCCGTGCTGAACGAGCTGGTGTCTGTGGAGGATCTGAAGAATTTTGAAAGGAAATTTCAGTCTGAGCAGGCAGCTGGTTCTGTGTCCAAGAGCACGCAATTTGAATATGCCTGGTGCCTGGTTCGAAGCAAATACAATGAGGACATCCGCAGAGGCATCGTGCTGCTGGAGGAGCTGTTGCCCAAAGGGAGCAAAGAGGAACAGCGGGACTATGTCTTCTACCTGGCCGTGGGCAACTACCGGCTCAAGGAATATGAAAAGGCTCTAAAGTATGTGCGAGGGCTGTTGCAGACTGAGCCCCAGAACAACCAGGCCAAGGAGCTGGAACGCCTGATTGATAAGGCCATGAAGAAAGATGGACTGGTAGGCATGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jichi Zhou et al.
Cell stress & chaperones, 24(2), 369-383 (2019-01-19)
Sirtuin 3 (Sirt3)-modified mitochondrial fission participates in the progression of several types of cancers. However, its role in tongue cancer requires investigation. The aim of our study is to determine whether Sirt3 knockdown regulates the viability of tongue cancer cells
Qinfang Shen et al.
Molecular biology of the cell, 25(1), 145-159 (2013-11-08)
Mitochondrial fission is mediated by the dynamin-related protein Drp1 in metazoans. Drp1 is recruited from the cytosol to mitochondria by the mitochondrial outer membrane protein Mff. A second mitochondrial outer membrane protein, named Fis1, was previously proposed as recruitment factor
Dongjoon Kim et al.
Cells, 9(7) (2020-07-16)
Diabetic retinopathy is a prevalent microvascular complication characterized by apoptotic vascular cell loss in the retina. Previous studies have shown that high glucose (HG)-induced mitochondrial fragmentation plays a critical role in promoting retinal vascular cell apoptosis. Here, we investigated whether
Mitsuo Kato et al.
Communications biology, 4(1), 30-30 (2021-01-06)
Diabetic kidney disease (DKD) is a major complication of diabetes. Expression of members of the microRNA (miRNA) miR-379 cluster is increased in DKD. miR-379, the most upstream 5'-miRNA in the cluster, functions in endoplasmic reticulum (ER) stress by targeting EDEM3.
Hidenori Otera et al.
The Journal of cell biology, 191(6), 1141-1158 (2010-12-15)
The cytoplasmic dynamin-related guanosine triphosphatase Drp1 is recruited to mitochondria and mediates mitochondrial fission. Although the mitochondrial outer membrane (MOM) protein Fis1 is thought to be a Drp1 receptor, this has not been confirmed. To analyze the mechanism of Drp1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service