Skip to Content
Merck
All Photos(1)

Key Documents

EMU011721

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Ovol1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGATCCCCAGAGCAGAGACCAGGGCTTTCTACGAACCAAGATGAAGGTGACCTTGGGGGACAGTCCAAATGGAGACCTTTTTACCTGCCACATCTGCCAGAAGTCCTTCACCCACCAGCGCATGCTGAATCGACACATGAAGTGTCACAACGACGTCAAGAGGCACCTCTGCACTTACTGTGGGAAGGGCTTCAATGACACCTTTGACCTCAAGAGACATGTCCGAACCCACACTGGTGTGCGGCCCTACAAGTGCAGTCTGTGTGATAAGGCCTTCACTCAGCGCTGCTCTCTGGAGTCCCACCTCAAGAAGATCCACGGAGTGCAGCAGAAGTATGCATACAAGGAGCGCCGGGCCAAGCTCTACGTGTGTGAGGAGTGCGGTTGCACATCCGAGAGTCAAGAGGGTCATGTCCTGCAC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen J Renaud et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(45), E6175-E6184 (2015-10-28)
Epithelial barrier integrity is dependent on progenitor cells that either divide to replenish themselves or differentiate into a specialized epithelium. This paradigm exists in human placenta, where cytotrophoblast cells either propagate or undergo a unique differentiation program: fusion into an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service