Skip to Content
Merck
All Photos(1)

Key Documents

EHU055701

Sigma-Aldrich

MISSION® esiRNA

targeting human NPY

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTCGCCCGACAGCATAGTACTTGCCGCCCAGCCACGCCCGCGCGCCAGCCACCATGCTAGGTAACAAGCGACTGGGGCTGTCCGGACTGACCCTCGCCCTGTCCCTGCTCGTGTGCCTGGGTGCGCTGGCCGAGGCGTACCCCTCCAAGCCGGACAACCCGGGCGAGGACGCACCAGCGGAGGACATGGCCAGATACTACTCGGCGCTGCGACACTACATCAACCTCATCACCAGGCAGAGATATGGAAAACGATCCAGCCCAGAGACACTGATTTCAGACCTCTTGATGAGAGAAAGCACAGAAAATGTTCCCAGAACTCGGCTTGAAGACCCTGCAATGTGGTGATGGGAAATGAGACTTGCTCTCTGGCCTTTTCCTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheni-Chery Sudhakumari et al.
General and comparative endocrinology, 251, 54-65 (2017-03-23)
Neuropeptide-Y (NPY) has diverse physiological functions which are extensively studied in vertebrates. However, regulatory role of NPY in relation to brain ontogeny and recrudescence with reference to reproduction is less understood in fish. Present report for the first time evaluated
Claire B de La Serre et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 311(5), R930-R939 (2016-11-03)
Increased neuropeptide Y (NPY) gene expression in the dorsomedial hypothalamus (DMH) has been shown to cause hyperphagia, but the pathway underlying this effect remains less clear. Hypothalamic neural systems play a key role in the control of food intake, in
Sung-Hyeok Hong et al.
Oncotarget, 6(9), 7151-7165 (2015-02-26)
Ewing sarcoma (ES) develops in bones or soft tissues of children and adolescents. The presence of bone metastases is one of the most adverse prognostic factors, yet the mechanisms governing their formation remain unclear. As a transcriptional target of EWS-FLI1
Min Hee Park et al.
The EMBO journal, 34(12), 1648-1660 (2015-04-29)
Many reports have revealed the importance of the sympathetic nervous system (SNS) in the control of the bone marrow environment. However, the specific role of neuropeptide Y (NPY) in this process has not been systematically studied. Here we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service