Skip to Content
Merck
All Photos(1)

Key Documents

Y0001514

Megestrol acetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

17α-Acetoxy-6-methyl-4,6-pregnadiene-3,20-dione, 17α-Hydroxy-6-methyl-4,6-pregnadiene-3,20-dione 17-acetate, 17α-Hydroxy-6-methyl-6-dehydroprogesterone 17-acetate, Megestrol 17-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
Beilstein:
1917291
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

megesterol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@]3(OC(C)=O)C(C)=O)[C@]1([H])C=C(C)C4=CC(=O)CC[C@]24C

InChI

1S/C24H32O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h12-13,18-20H,6-11H2,1-5H3/t18-,19+,20+,22-,23+,24+/m1/s1

InChI key

RQZAXGRLVPAYTJ-GQFGMJRRSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Megestrol acetate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yo Han Kim et al.
Clinical therapeutics, 37(2), 439-447 (2014-12-03)
Megestrol acetate oral suspension is an appetite stimulant indicated for cachexia. It is available in a conventional formulation and as a nanocrystal dispersion. The aim of this study was to compare the tolerability and pharmacokinetics of these formulations under fed
Navdha Mittal et al.
PloS one, 9(10), e110410-e110410 (2014-10-24)
Resistance to progestin treatment is a major hurdle in the treatment of advanced and reoccurring endometrial cancer. Fenretinide is a synthetic retinoid that has been evaluated in clinical trials as a cancer therapeutic and chemo-preventive agent. Fenretinide has been established

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service