Skip to Content
Merck
All Photos(1)

Key Documents

W420555

Sigma-Aldrich

Prenyl formate

≥95%, FG

Synonym(s):

3-methylbut-2-enyl formate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
FEMA Number:
4205
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.694
NACRES:
NA.21

grade

FG

Quality Level

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

application(s)

flavors and fragrances

food allergen

no known allergens

Organoleptic

rum; tomato; fruity; ethereal

SMILES string

C\C(C)=C\COC=O

InChI

1S/C6H10O2/c1-6(2)3-4-8-5-7/h3,5H,4H2,1-2H3

InChI key

ZCROFVOAWLRGFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service