Skip to Content
Merck
All Photos(3)

Key Documents

M2545

Sigma-Aldrich

Magnesium acetate tetrahydrate

BioXtra, ≥99%

Synonym(s):

Acetic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Mg · 4H2O
CAS Number:
Molecular Weight:
214.45
Beilstein:
3730605
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39021909
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

product line

BioXtra

Quality Level

Assay

≥99%

form

solid

impurities

≤0.0005% Phosphorus (P)
≤0.005% Insoluble matter

mp

72-75 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.002%
Ba: ≤0.001%
Ca: ≤0.002%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mn: ≤0.005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Sr: ≤0.005%
Zn: ≤0.001%

SMILES string

O.O.O.O.CC(=O)O[Mg]OC(C)=O

InChI

1S/2C2H4O2.Mg.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

XKPKPGCRSHFTKM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis and application of SiO2-MgO coated MWCNTs: Magnesium acetate tetrahydrate was utilized in the synthesis of SiO2-MgO coatings on multi-walled carbon nanotubes (MWCNTs). This method showcases the versatility of magnesium acetate as a precursor in creating advanced materials for potential applications in molecular biology and biochemical research, enhancing the performance of functional materials in various scientific assays (Nemeth et al., 2019).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christiane Iserman et al.
Cell, 181(4), 818-831 (2020-05-04)
Cells sense elevated temperatures and mount an adaptive heat shock response that involves changes in gene expression, but the underlying mechanisms, particularly on the level of translation, remain unknown. Here we report that, in budding yeast, the essential translation initiation
Yoshihito Kitaoku et al.
Planta, 242(4), 895-907 (2015-05-23)
We first solved the crystal structure of class III catalytic domain of a chitinase from fern (PrChiA-cat), and found a structural difference between PrChiA-cat and hevamine. PrChiA-cat was found to have reduced affinities to chitin oligosaccharides and allosamidin. Plant class
Catarina M S S Neves et al.
Physical chemistry chemical physics : PCCP, 17(47), 32044-32052 (2015-11-18)
Due to scarce available experimental data, as well as due to the absence of predictive models, the influence of salts on the solubility of ionic liquids (ILs) in water is still poorly understood. To this end, this work addresses the
Andrey G Malyutin et al.
ACS applied materials & interfaces, 7(22), 12089-12098 (2015-05-20)
Here we explore the formation of virus-like nanoparticles (VNPs) utilizing 22-24 nm iron oxide nanoparticles (NPs) as cores and proteins derived from viral capsids of brome mosaic virus (BMV) or hepatitis B virus (HBV) as shells. To accomplish that, hydrophobic
Tanja Rozman Peterka et al.
Journal of pharmaceutical and biomedical analysis, 110, 67-75 (2015-03-26)
Tacrolimus is macrolide drug that is widely used as a potent immunosuppressant. In the present work compatibility testing was conducted on physical mixtures of tacrolimus with excipients and on compatibility mixtures prepared by the simulation of manufacturing process used for

Questions

  1. Would you please provide stability data for this material?

    1 answer
    1. This product is quite stable when stored at the recommended -20 °C. The material is not assigned an expiration date, but it has a recommended retest date of 4 years from the date of quality release. The compound is moisture sensitive and should be stored tightly sealed at ambient temperature.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service