Skip to Content
Merck
All Photos(2)

Key Documents

63067

Millipore

Magnesium citrate tribasic nonahydrate

suitable for microbiology, ≥95.0% (KT)

Synonym(s):

Citric acid magnesium salt, Trimagnesium dicitrate nonahydrate, tri-Magnesium dicitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg3(C6H5O7)2 · 9H2O
CAS Number:
Molecular Weight:
613.25
Beilstein:
6674777
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

sterility

non-sterile

Quality Level

Assay

≥95.0% (KT)

form

powder

anion traces

chloride (Cl-): ≤70 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤500 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤500 mg/kg
Na: ≤200 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

application(s)

microbiology

SMILES string

O.O.O.O.O.O.O.O.O.OC2(CC(=O)O[Mg]OC(=O)CC1(O)CC(=O)O[Mg]OC1=O)CC(=O)O[Mg]OC2=O

InChI

1S/2C6H8O7.3Mg.9H2O/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;;;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;9*1H2/q;;3*+2;;;;;;;;;/p-6

InChI key

IYQJAGXFXWIEJE-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonia E Schantl et al.
Nature communications, 11(1), 721-721 (2020-02-07)
Myo-inositol hexakisphosphate (IP6) is a natural product known to inhibit vascular calcification (VC), but with limited potency and low plasma exposure following bolus administration. Here we report the design of a series of inositol phosphate analogs as crystallization inhibitors, among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service