Skip to Content
Merck
All Photos(2)

Key Documents

11667289001

Roche

Sodium Dodecyl Sulfate (SDS)

anionic, electrophoresis grade

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium Dodecyl Sulfate (SDS), Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25NaO4S
CAS Number:
Molecular Weight:
288.38
MDL number:
UNSPSC Code:
12161900

Assay

≥97% (GC)

form

powder

mol wt

288.38 g/mol

packaging

pkg of 1 kg

manufacturer/tradename

Roche

technique(s)

HPLC: suitable
LC/MS: suitable

solubility

H2O: soluble (clear solution (100 mg/ml in H2O))

absorption

≤0.1 at 280 nm in water at 30 mg/mL

shipped in

ambient

storage temp.

20-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])OCCCCCCCCCCCC

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

General description

Pure quality, particularly suited for SDS-gel electrophoresis.

Application

Suitable in most routine electrophoretic applications as a denaturing agent for proteins.

Quality

Purity: ≥99.5 alkyl sulfate, ≥97% dodecyl sulfate (GC)
Contaminants: ≤5 ppm Pb (AA), ≤0.5% Cl (AA)

C12 fatty acids: >97% (after saponification)
Ease of removal*: –
Critical micellar concentration: 8.3 mM at +25°C
Formula: C12H25NaSO4

Preparation Note

Working concentration: > 10 mg detergent per mg protein

Other Notes

For life science research only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 2 Oral - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service