Skip to Content
Merck
All Photos(1)

Documents

MABS2253

Sigma-Aldrich

Anti-LRP1 α-chain Antibody, clone 8G1

Synonym(s):

Anti-LRP1 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8G1, monoclonal

mol wt

calculated mol wt 504.61 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

General description

Prolow-density lipoprotein receptor-related protein 1 (UniProt Q07954; also known as A2MR, Alpha-2-macroglobulin receptor, APER, Apolipoprotein E receptor, CD19. LRP-1) is encoded by the LRP1 (also known as A2MR, APR) gene (Gene ID: 4035) in human. LRP-1 is single pass type I membrane protein that is expressed in most tissues, but is abundant in liver, brain, and lung. It serves as a major VSMC receptor with a crucial role in maintaining vessel wall integrity and its expression in adult smooth muscle cells is shown to be essential for maintaining a contractile phenotype. LRP1 is synthesized with a signal peptide sequence (aa 1-19) and is processed in trans-Golgi network by Furin to generate a 515 kDa a subunit and an 85 kDa b subunit that contains the 100-amino intracellular domain (LRPICD; aa 4445-4544). The a and b subunits are non-covalently linked during LRP1 transport to the cell membrane. LRP1 recognizes and mediates the endocytosis of more than 40 different ligands, including apolipoprotein E (ApoE), APP and amyloid b. It is the primary receptor mediating transport of amyloid b peptides across the blood-brain barrier into circulation, thereby clearing them from the brain. LRP1 is required for early embryonic development and is involved in cellular lipid homeostasis and plasma clearance of chylomicron remnants and activated LRPAP1 (a2-macroglobulin). Excessive copper accumulation in the brain has been linked with reduced LRP1 mediated clearance of amyloid b peptides across the blood brain barrier, which may contribute to complications of Alzheimer′s disease.

Specificity

Clone 8G1 is a mouse monoclonal antibody that detects human LRP-1. It targets an epitope within the a chain in the extracellular domain.

Immunogen

Full-length LRP-1 purified from human placenta and 515 kDa band excised from gel and used as immunogen.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in MCF-7 cells.

Immunocytochemistry Analysis: A 1:50 dilution of this antibody detected LRP1 a-chain in MCF-7 cells.

Tested Applications

Immunocytochemistry Analysis: A representative lot detected LRP1 -chain in Immunocytochemistry applications (Craig, J., et al. (2013). PLoS One. 8(7):e70432).

Immunoprecipitation Analysis: A representative lot immunoprecipitated LRP1 -chain in Immunoprecipitation applications (Ranganathan, S., et al. (2011). J. Biol. Chem.286(35):30535-30541; Boye, K., et al. (2017). Nat. Commun. 8(1):1571).

Western Blotting Analysis: A representative lot detected LRP1 -chain in Western Blotting applications (Strickland, D.K., et al. (1990). J Biol Chem.;265(29):17401-4; Yamamoto, K., et al. (2013). FASEB J.;27(2):511-21; Yamamoto, K. et al. (2016). Matrix Biol.;56:57-73; Boye, K., et al. (2017). Nat Commun.;8(1):1571).

ELISA Analysis: A representative lot detected LRP1 -chain in ELISA applications (Ranganathan, S., et al. (2011). J. Biol. Chem. 286(35):30535-3054).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-LRP1 a-chain, clone 8G1, Cat. No. MABS2253, is a mouse monoclonal antibody that detects LRP1 a -chain and is tested for use in ELISA, Immunocytochemistry, Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service