Skip to Content
Merck
All Photos(1)

Key Documents

MAB4603

Sigma-Aldrich

Anti-Caspase 3 Antibody, large subunit & proform, clone 3CSP03

clone 3CSP03, Chemicon®, from mouse

Synonym(s):

CPP32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3CSP03, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP3(836)

Specificity

Recognizes a protein of 32 kDa, identified as the Caspase-3 protein. Highly specific to Caspase-3 and shows no cross-reaction with other caspase family members. Caspase-3 is ubiquitously expressed and like other caspases is synthesized as an inactive proenzyme. Upon activation, caspase-3 is cleaved generating two subunits of 17 kDa and 12 kDa respectively. Caspase-3 has been implicated in the induction of apoptosis. Activation of caspase-3 occurs in response to a variety of apoptotic inducers, including Fas-mediated apoptosis.

Immunogen

Epitope: large subunit & proform
Recombinant full-length human caspase-3 protein

Application

Detect Caspase 3 using this Anti-Caspase 3 Antibody, large subunit & proform, clone 3CSP03 validated for use in IP, WB, IH(P).
Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Caspases

Enzymes & Biochemistry
Western blot: 1 μg/ml for 2 hours at RT.

Immunoprecipitation: 2 μg/mg of protein lysate Immunohistochemistry (frozen and paraffin): 2-4 μg/ml for 30 minutes at RT. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6.0, for 10-20 min. followed by cooling for 20 min. at RT.

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-440; 04-1088

Physical form

Format: Purified
Liquid in 10 mM PBS, pH 7.4, with 0.2% BSA and 15 mM sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaposi's sarcoma herpesvirus microRNAs target caspase 3 and regulate apoptosis.
Suffert, G; Malterer, G; Hausser, J; Viiliainen, J; Fender, A; Contrant, M; Ivacevic et al.
PLoS Pathogens null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service