Skip to Content
Merck
All Photos(2)

Key Documents

EZFITL0W06

Millipore

EZ-Fit® Manifold, 6-place for Microfil® V or Microfil® S filtration units

General microbiological analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

stainless steel

Sterilization Compatibility

autoclavable compatible

manufacturer/tradename

EZ-Fit®

input

sample type pharmaceutical(s)
sample type bottled water
sample type beer
sample type cosmetics

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

General description

EZ-Fit Manifold is available with the new Microfil head for use with presterilized, ready-to-use funnels and membranes.

Application

General microbiological analysis

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service