Skip to Content
Merck

ABD70

Anti-Fbxl16 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

F-box/LRR-repeat protein 16, F-box and leucine-rich repeat protein 16

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Fbxl16, also known as F-box/LRR repeat protein 16, F-box and leucine-rich repeat protein 16, and encoded by the gene Fbxl16, is the protein responsible for the substrate recognition of SCF. SCF is the SKP1-CUL1-F-box protein- type E3 ubiquitin ligase complex. The Skp1-Cul1-F-box protein (SCF) complex is one of the most well-characterized types of ubiquitin ligase E3 proteins, with the E3 activity of the complex being regulated in part at the level of complex formation. The complex has important roles in the ubiquitination of proteins involved in the cell cycle and also marks various other cellular proteins for destruction. Fbxl16 appears to be a novel E2F1-regulated gene whose expression can be up regulated when particular cell cycle proteins are disrupted, and thus Fbxl16 expression is often high in cancerous tumors.
~52 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Immunogen

KLH-conjugated linear peptide corresponding to mouse Fbxl16.

Application

Anti-Fbxl16 Antibody detects level of Fbxl16 & has been published & validated for use in Fbxl16.
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected Fbxl16 in human cerebral cortex and thalamus tissues.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Fbxl16 in human ileum tissues.

Western Blotting Analysis: 1 µg/mL of this antibody detected Fbxl16 in NIH/3T3 cell lysates, which were untransfected, transfected with Myc-tagged Fbxl16, and transfected with a negative control siRNA, and not in NIH/3T3 cell lysate transfected with Fbxl16 siRNA (Prof. Ray De Shaies, California Institute of Technology, Pasadena.).
Research Category
Stem Cell Research
Research Sub Category
Apoptosis - Additional

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in serum starved HeLa cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Fbxl16 in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library