Skip to Content
Merck
All Photos(1)

Documents

8.41636

Sigma-Aldrich

2-Aminoethyldiphenyl borate

for synthesis

Synonym(s):

2-Aminoethyldiphenyl borate, Diphenylboric acid-2-aminoethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16BNO
CAS Number:
Molecular Weight:
225.09
MDL number:
UNSPSC Code:
12352116
EC Index Number:
208-366-5
NACRES:
NA.22

Quality Level

form

powder

mp

189-194 °C

InChI

1S/C14H16BNO/c16-11-12-17-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10H,11-12,16H2

InChI key

BLZVCIGGICSWIG-UHFFFAOYSA-N

Application

  • Investigating the Potentiation Effect of 2-APB on CRAC Channels: This study focused on the dual actions of 2-APB, which can potentiate and inhibit calcium influx via CRAC channels, aiming to unravel the underlying mechanisms (X Xu, SA Syed, T Xu - Biophysical Journal, 2015).
  • Inhibitory Effects of 2-Aminoethoxydiphenyl Borate (2-APB) on Three KV1 Channel Currents: Explores 2-APB′s role as a multitarget compound with potential as a drug precursor, highlighting its effects on KV1 channel currents (W Zhao, et al. - Molecules, 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service