8.02358
Isobutyl chloroformate
for synthesis
Synonym(s):
Isobutyl chloroformate, Chloroformic acid isobutyl ester
About This Item
Recommended Products
vapor pressure
12 hPa ( 20 °C)
Quality Level
Assay
≥95.0% (GC)
form
liquid
potency
1400 mg/kg LD50, oral (Rat)
reaction suitability
reaction type: Coupling Reactions
transition temp
flash point 30 °C
density
1.05 g/cm3 at 20 °C
application(s)
peptide synthesis
storage temp.
15-25°C
SMILES string
ClC(=O)OCC(C)C
InChI
1S/C5H9ClO2/c1-4(2)3-8-5(6)7/h4H,3H2,1-2H3
InChI key
YOETUEMZNOLGDB-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.043 - 1.046
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
96.8 °F - closed cup
Flash Point(C)
36 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service