Skip to Content
Merck
All Photos(1)

Key Documents

5.89571

Sigma-Aldrich

1,2-Dichlorobenzene

99%, anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein:
606078
MDL number:
UNSPSC Code:
12191501
NACRES:
NA.22
grade:
anhydrous
Assay:
99%
bp:
178-180 °C (lit.)
vapor pressure:
1.2 mmHg ( 20 °C)
1.6 mmHg ( 35 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

5.1 (vs air)

vapor pressure

1.2 mmHg ( 20 °C)
1.6 mmHg ( 35 °C)

Assay

99%

autoignition temp.

1198 °F

expl. lim.

9.2 %

refractive index

n20/D 1.551 (lit.)

bp

178-180 °C (lit.)

mp

−18-−17 °C (lit.)

density

1.306 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1Cl

InChI

1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

RFFLAFLAYFXFSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service