Skip to Content
Merck
All Photos(2)

Key Documents

880157P

Avanti

DSPE-PEG2K-triGalNAc

Avanti Research - A Croda Brand

Synonym(s):

DSPE-PEG2K-triGalNAc, 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[carboxy(polyethylene glycol)-2000]-N-tri(N-acetylgalactosamine), ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C183H355N10O81P
CAS Number:
Molecular Weight:
4022.77
UNSPSC Code:
12352211
NACRES:
NA.26

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (880157P-1MG)
pkg of 1 × 10 mg (880157P-10MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

shipped in

dry ice

storage temp.

−20°C

Application

drug delivery

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service