Skip to Content
Merck
All Photos(1)

Key Documents

870850P

Avanti

C12 Carnitine

Avanti Research - A Croda Brand 870850P, powder

Synonym(s):

lauroyl L-carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H37NO4
CAS Number:
Molecular Weight:
343.50
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (870850P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870850P

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCCCCCC)=O)CC([O-])=O

InChI

1S/C19H37NO4/c1-5-6-7-8-9-10-11-12-13-14-19(23)24-17(15-18(21)22)16-20(2,3)4/h17H,5-16H2,1-4H3/t17-/m1/s1

InChI key

FUJLYHJROOYKRA-QGZVFWFLSA-N

General description

C12 Carnitine or lauroyl L-carnitine, is a lauric acid-derived serum acylcarnitine. It mediates obesity-induced inflammation in adipose. Elevated levels of lauroyl carnitine in carcinoma-associated fibroblasts (CAF)-adipose tissue acts as a biomarker for obesity, insulin resistance and inflammation.

Application

C12 Carnitine or lauroyl L-carnitine has been used to investigate its effect on the glycine transporters activity. It has also been used to study its effect on pro-inflammatory activation of macrophages.

Packaging

5 mL Amber Glass Screw Cap Vial (870850P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brante P Sampey et al.
PloS one, 7(6), e38812-e38812 (2012-06-16)
Obesity has reached epidemic proportions worldwide. Several animal models of obesity exist, but studies are lacking that compare traditional lard-based high fat diets (HFD) to "Cafeteria diets" (CAF) consisting of nutrient poor human junk food. Our previous work demonstrated the
Oleoyl-l-carnitine inhibits glycine transport by G ly T 2
Carland JE, et al.
British Journal of Pharmacology, 168(4), 891-902 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service