Skip to Content
Merck
All Photos(2)

Documents

840036P

Avanti

10:0 PS

1,2-didecanoyl-sn-glycero-3-phospho-L-serine (sodium salt), powder

Synonym(s):

PS(10:0/10:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H49NO10PNa
CAS Number:
Molecular Weight:
589.63
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840036P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840036P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCCCC)=O)COC(CCCCCCCCC)=O.[Na+]

InChI

1S/C26H50NO10P.Na/c1-3-5-7-9-11-13-15-17-24(28)34-19-22(20-35-38(32,33)36-21-23(27)26(30)31)37-25(29)18-16-14-12-10-8-6-4-2;/h22-23H,3-21,27H2,1-2H3,(H,30,31)(H,32,33);/q;+1/p-1/t22-,23+;/m1./s1

InChI key

MNHRZRUSKRPBDA-RFPXDPOKSA-M

General description

Glycerophosphoserines (PS) associates with spectrin and maintains the membrane asymmetry of red blood cells (RBC).

Application

1,2-didecanoyl-sn-glycero-3-phospho-L-serine has been used as a component in internal lipid standard mixture for matrix-assisted laser desorption/ionization (MALDI)-fourier-transform ion cyclotron resonance (FTICR)-mass spectrometry (MS) analysis.

Packaging

5 mL Clear Glass Sealed Ampule (840036P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phospholipidomics reveals differences in glycerophosphoserine profiles of hypothermically stored red blood cells and microvesicles
Bicalho B, et al.
Biochimica et Biophysica Acta - Biomembranes, 1828(2), 317-326 (2013)
Changes in lipid distribution in E. coli strains in response to norfloxacin
Schenk ER, et al.
Journal of Mass Spectrometry : Jms, 50(1), 88-94 (2015)
Emily R Schenk et al.
Journal of mass spectrometry : JMS, 50(1), 88-94 (2015-01-21)
Bacterial resistance to antibiotics has become an increasing threat, requiring not only the development of new targets in drug discovery, but more importantly, a better understanding of cellular response. In the current study, three closely related Escherichia coli strains, a
Peter A Leventis et al.
Annual review of biophysics, 39, 407-427 (2010-03-03)
Phosphatidylserine (PS) is the most abundant negatively charged phospholipid in eukaryotic membranes. PS directs the binding of proteins that bear C2 or gamma-carboxyglutamic domains and contributes to the electrostatic association of polycationic ligands with cellular membranes. Rather than being evenly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service