Key Documents
Z740555
Sigma-Aldrich
Aldrich® Essentials beaker, Griffin
low form, capacity 10 mL, ungraduated
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
feature
graduations: no
packaging
pkg of 48 ea
manufacturer/tradename
Sigma-Aldrich
capacity
10 mL
Related Categories
General description
Borosilicate glass, Griffin style, low form beakers with reference graduations and pour spout. Uniform, standard walls and flat bottom for even heating. Meet ASTM E960 designation.
Other Notes
Browse all glass beakers
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)