Key Documents
Z549711
Sigma-Aldrich
High-vacuum valve adapter
male joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Pricing and availability is not currently available.
Joint
male joint: ST/NS29/32
General description
For high-vacuum applications. Features a high-vacuum PTFE valve. For use with 1/4in. I.D. tubing.
Choose from one of the most recent versions:
Certificates of Analysis (COA)