Skip to Content
Merck
All Photos(1)

Key Documents

Z162965

Zipper-top poly bag

size 5 in. × 8 in.

Synonym(s):

Ziplock Storage Bag, Zippered Storage Bag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24111500
NACRES:
NB.35
Pricing and availability is not currently available.

material

clear low-density polyethylene

Agency

suitable for EPA OTM-45

size

5 in. × 8 in.

thickness

4 mil

application(s)

PFAS testing

Looking for similar products? Visit Product Comparison Guide

General description

Zipper-top poly bag is see-through with a double-track zipper. It meets FDA and USDA specifications for food contact. The bags also have a helpful hang hole at the top.

Application

The zipper-top poly bag has been used to store marine sediment samples.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel L Coppock et al.
Environmental pollution (Barking, Essex : 1987), 230, 829-837 (2017-07-25)
Microplastics (plastic particles, 0.1 μm-5 mm in size) are widespread marine pollutants, accumulating in benthic sediments and shorelines the world over. To gain a clearer understanding of microplastic availability to marine life, and the risks they pose to the health

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service