Skip to Content
Merck
All Photos(1)

Key Documents

W242101

Sigma-Aldrich

Ethyl 2-aminobenzoate

≥96%, FCC, FG

Synonym(s):

Ethyl anthranilate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2C2H5
CAS Number:
Molecular Weight:
165.19
FEMA Number:
2421
Beilstein:
878874
EC Number:
Council of Europe no.:
251
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.716
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

5.7 (vs air)

Assay

≥96%

refractive index

n20/D 1.564 (lit.)

bp

129-130 °C/9 mmHg (lit.)

mp

13-15 °C (lit.)

density

1.117 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

blossom; grape; sweet

SMILES string

CCOC(=O)c1ccccc1N

InChI

1S/C9H11NO2/c1-2-12-9(11)7-5-3-4-6-8(7)10/h3-6H,2,10H2,1H3

InChI key

TWLLPUMZVVGILS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoying Sun et al.
Molecular vision, 17, 2191-2196 (2011-08-26)
Pirfenidone (5-methyl-1-phenyl-2-[1H]-pyridone) is a new, broad-spectrum agent that has an inhibition effect on the proliferation, migration, and collagen contraction of human Tenon's fibroblasts, and thus modulating the wound healing process of glaucoma filtering surgical site. This study investigated the pharmacokinetics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service