Key Documents
SYNC351922
Synthware reflux condenser
joint: ST/NS 19/22, coil L 100 mm
Synonym(s):
Synthware C351922, coil-type condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C351922
coil L
100 mm
Related Categories
General description
Coil section is 100 mm in height.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)