SLW1020/06D
Pyrex® Beakers, conical form, Phillips pattern
250 mL
Synonym(s):
Pyrex, SciLabware 1020/06D
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
1020/06D
SciLabware
H
105 mm
base diam.
68.0 mm
top diam.
52 mm
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Conical shape (Phillips pattern) with spout
- Manufactured from ′Pyrex borosilicate glass
- Excellent thermal performance and chemical resistance
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service