Key Documents
MT30207958
Mettler Toledo Seven2Go DO meter S4
Meter Only, polarographic
About This Item
UNSPSC Code:
41115611
NACRES:
NB.61
Pricing and availability is not currently available.
description
connector BNC/RCA
pH/ion/ORP meters
feature
Meter Only, polarographic
manufacturer/tradename
Mettler-Toledo 30207958
dissolved oxygen measuring range
0.00-99.9 mg/L (ppm)
pressure measuring range
500-1100 (mbar)
saturation measuring range
0.0-600% (DO)
temperature measuring range
0-60 °C
battery power
4 x 1.5 V AA (or 1.3 V NiMH accumulators)
General description
Simple and reliable, attractively designed and robust. The Seven2Go portables are engineered to provide users with fast quality data, one-handed operation, and durability, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors.
Simple Menu – Intuitive Operation:
  • Seven2Go′s new intuitive menu guarantees out-of-the-box operation for anyone. Users no longer need to study lengthy operating instructions
T-Pad – Comfortable, Fast Navigation:
  • Seven2Go′s new T-Pad improves the speed with which you move through menus. The smart button arrangement allows for faster navigation that provide you shorter time to actual measurement
ISM® Ready – Reduced Chances for Error:
  • Upon connection of any InLab ISM (Intelligent Sensor Management) sensor, the instrument automatically recognizes the sensor and stores sensor ID, serial number, and last calibration data
IP67 – Weatherproof and Robust Design:
  • Thanks to its IP67 protection and drop test resistance, the new Seven2Go portables are able to withstand harsh and demanding environments
Features and Benefits
Includes: Meter, USB cable, electrode clip, base stand, instructions, quick guide, test certificate, declaration of conformity, batteries
Legal Information
ISM is a registered trademark of Mettler-Toledo, Inc.
Seven2Go is a trademark of Mettler-Toledo, Inc.
  • Choose from one of the most recent versions:
    Certificates of Analysis (COA)