Skip to Content
Merck
All Photos(1)

Key Documents

GF08037592

Glass Sealing Alloy, Fe54Ni29Co17

100mm tube, 2.0mm outside diameter, 0.5mm wall thickness, 1.0mm inside diameter, as drawn

Synonym(s):

FE047301

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 080-375-92

L × wall thickness

100 mm × 0.5 mm

O.D. × I.D.

2.0 mm × 1.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service