DWKBWP0250P
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap
round translucent polypropylene bottle, capacity 250 mL
Synonym(s):
BWP0250P
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
round translucent polypropylene bottle
wide-neck
packaging
pkg of 10 ea
manufacturer/tradename
Azlon BWP0250P
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Featuring a leakproof, linerless cap design, AZLON® wide neck polypropylene bottles are easy to fill and autoclavable at 121°C.
Application
Ideal for a wide range of liquid storage and sampling applications
Features and Benefits
Autoclavable at 121°C and suitable for food contact applications
Legal Information
AZLON is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service