Skip to Content
Merck
All Photos(2)

Key Documents

DWKBGF130P

Azlon Wash Bottles with Integral Spout

low-density polyethylene bottle, capacity 250 mL

Synonym(s):

BGF130P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41000000
NACRES:
NB.43

material

low-density polyethylene bottle

packaging

pkg of 5 ea

manufacturer/tradename

Azlon BGF130P

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

General description

Featuring an ergonomic design, this AZLON® wash bottle with integral spout has an integrally moulded dispensing tube which minimises contamination events that occur during filling.

Application

Ideal for dispensing liquids

Features and Benefits

Rectangular design specifically suited for smaller hands

Legal Information

AZLON is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service