BAH500250546
SP Bel-Art® Glove Box Economy Sleeved Gloves for 8 in. Glove Ports
size 10 large
Synonym(s):
glove box
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
Recommended Products
material
black neoprene
size
10 large
8 in. (Glove port )
packaging
pkg of 1 pair
manufacturer/tradename
Bel-Art H50025-0546
Related Categories
General description
One piece, bonded neoprene protection against liquid or gas penetration.
Features and Benefits
- Use series 50029 for glove boxes with 6” glove ports (F50030-0000, F50028-2000)
- Use series 50025 for glove boxes with 8” glove ports (H50026-0000)
- Requires clamping rings to hold gloves in place
- Sold in pairs
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)