903809
Cholic-2,2,4,4-d4 acid 3-sulfate disodium salt
≥98 atom % D, ≥98% (CP)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C24D4H34Na2O8S
Molecular Weight:
536.62
UNSPSC Code:
12352106
NACRES:
NA.24
Recommended Products
isotopic purity
≥98 atom % D
Quality Level
Assay
≥98% (CP)
form
powder
technique(s)
mass spectrometry (MS): suitable
mass shift
M+4
storage temp.
−20°C
SMILES string
O=C(O[Na])CC[C@H]([C@@H](CC1)[C@]([C@@H](O)C2)(C)C1C([C@H](O)C3)C2[C@]4(C)[C@@]3([H])C([2H])([2H])[C@H](OS(=O)(O[Na])=O)C([2H])([2H])C4)C
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service